Learn More
Exemestane, MedChemExpress
MedChemExpress Exemestane (FCE 24304) is a selective, irreversible and orally active steroidal aromatase inhibitor with IC50s of 30 nM and 40 nM for human placental and rat ovarian aromatase, respectively. Exemestane can be used for hormone-dependent breast cancer research.
Brand: Medchem Express HY-13632
Description
Exemestane (FCE 24304) is a selective, irreversible and orally active steroidal aromatase inhibitor with IC50s of 30 nM and 40 nM for human placental and rat ovarian aromatase, respectively. Exemestane can be used for hormone-dependent breast cancer research.Specifications
Exemestane | |
107868-30-4 | |
10 mg | |
C20H24O2 | |
FCE 24304 EXE | |
C[C@]1([C@](CC2)([H])[C@]3([H])CC(C4=CC(C=C[C@]4(C)[C@@]3([H])CC1)=O)=C)C2=O | |
296.4 | |
Research |
Solid | |
White | |
Research | |
Cancer-programmed cell death | |
DMSO : ≥ 54 mg/mL (182.19 mM) | |
296.4 | |
99.92% |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only