missing translation for 'onlineSavingsMsg'
Learn More

Promed Bioscience Atelocollagen, Porcine Type I (Enzymatic extraction)

Lyophilized, highly purified Type I atelocollagen with superior functionality and physical/mechanical properties, ideal for cell/tissue culture and cell functional assays

Brand:  Promed Bioscience PRO-2001-20

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 17582835

  • 1469.21 SEK / 20mg
Estimated Shipment: 06-09-2024
to see stock.

Description

Description

  • Lyophilized, highly purified Type I atelocollagen with superior functionality and physical/mechanical properties (such as tensile strength and elasticity)
  • Proprietary enzyme-based extraction removes telopeptides at the non-helical N and C termini, resulting in a triple helix configuration that exhibits reduced immunogenicity and enhanced biocompatibility
  • Suitable for in vitro cell cultures, for cell functional assays, as a chemoattractant agent and for soft 3D cell cultures (e.g. tumor assays)
Specifications

Specifications

Lyophilized Fibrous Powder
20 mg
<1.0 EU/mL
Dialysis
-20°C to 8°C
>95% (by SDS/Western blotting)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers