missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promed Bioscience Atelocollagen, Porcine Type I (Enzymatic extraction)
Lyophilized, highly purified Type I atelocollagen with superior functionality and physical/mechanical properties, ideal for cell/tissue culture and cell functional assays
929.50 SEK - 1469.21 SEK
Specifications
Formulation | Lyophilized Fibrous Powder |
---|---|
Purification Method | Dialysis |
Content And Storage | -20°C to 8°C |
Endotoxin Level | <1.0 EU/mL |
Purity or Quality Grade | >95% (by SDS/Western blotting) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17572835
|
Promed Bioscience
PRO-2001-10 |
10 mg |
929.50 SEK
10mg |
Estimated Shipment: 06-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17582835
|
Promed Bioscience
PRO-2001-20 |
20 mg |
1469.21 SEK
20mg |
Estimated Shipment: 06-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Lyophilized, highly purified Type I atelocollagen with superior functionality and physical/mechanical properties (such as tensile strength and elasticity)
- Proprietary enzyme-based extraction removes telopeptides at the non-helical N and C termini, resulting in a triple helix configuration that exhibits reduced immunogenicity and enhanced biocompatibility
- Suitable for in vitro cell cultures, for cell functional assays, as a chemoattractant agent and for soft 3D cell cultures (e.g. tumor assays)
Specifications
Lyophilized Fibrous Powder | |
-20°C to 8°C | |
>95% (by SDS/Western blotting) |
Dialysis | |
<1.0 EU/mL |