missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX ATR/ATRIP Protein, Active

Active, Recombinant, human FLAG-tagged ATR full length (GenBank NM_001184.3*) and recombinant, human cMyc-tagged ATRIP full length, for use in Kinase Assays

10266.62 SEK - 88942.51 SEK

Specifications

Formulation mg/mL of enzyme in storage buffer.
Molecular Weight (g/mol) 303 kDa (ATR) and 87 kDa (ATRIP)
Species Human
Recombinant Recombinant
Protein Tag ATR-FLAG-tag / ATRIP-cMyc-tag
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17360844
View Documents Promotion Details
Eurofins DiscoverX
14-953
10 μg
10266.62 SEK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 03-09-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
17370844
View Documents Promotion Details
Eurofins DiscoverX
14-953M
250 μg
88942.51 SEK
250µg
View alternative products
Save Up to 
»
Estimated Shipment: 26-09-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Serine/threonine-protein kinase ATR activates checkpoint signaling upon genotoxic stresses such as ionizing radiation (IR), ultraviolet light (UV), or DNA replication stalling, thereby acting as a DNA damage sensor. ATR interacts with the ataxia telangiectasia and Rad3 related protein kinase, resulting in its accumulation at intranuclear foci induced by DNA damage.

GenBank NM_001184.3* / GenBank AF451323.1 * Note: the recombinant ATR contains the amino acid substitution M211T with reference toGenBank NM_001184.3. M211T is reported inGenBank XM_001131387.1.

Specifications

Specifications

mg/mL of enzyme in storage buffer.
Human
ATR-FLAG-tag / ATRIP-cMyc-tag
Full-length
ATR/ATRIP, active
303 kDa (ATR) and 87 kDa (ATRIP)
Recombinant
Recombinant enzyme expressed in mammalian cells
Combined ≥43% ration 1:1 to 2:1 ATR:ATRIP
Videos
SDS
Documents

Documents

Product Certifications