missing translation for 'onlineSavingsMsg'
Learn More

Avadomide, MedChemExpress

MedChemExpress Avadomide (CC 122) is an orally active cereblon modulator. Avadomide modulates cereblon E3 ligase activity and induces apoptosis of diffuse large B-cell lymphoma (DLBCL) cell lines. Avadomide exhibits potent antitumor and immunomodulatory activities.

Encompass_Preferred

513.50 SEK - 4014.63 SEK

Specifications

Chemical Name or Material Avadomide
CAS 1015474-32-4
Color Gray
Purity Grade Notes Research
Molecular Formula C14H14N4O3
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16475787
Encompass_Preferred
View Documents
Medchem Express
HY-100507
5 mg
513.50 SEK
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16459616
Encompass_Preferred
View Documents
Medchem Express
HY-100507
50 mg
2489.70 SEK
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16403058
Encompass_Preferred
View Documents
Medchem Express
HY-100507
10 mg
809.15 SEK
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16424356
Encompass_Preferred
View Documents
Medchem Express
HY-100507
100 mg
4014.63 SEK
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Avadomide (CC 122) is an orally active cereblon modulator. Avadomide modulates cereblon E3 ligase activity and induces apoptosis of diffuse large B-cell lymphoma (DLBCL) cell lines. Avadomide exhibits potent antitumor and immunomodulatory activities.
Specifications

Specifications

Avadomide
Gray
C14H14N4O3
DMSO : 33.33 mg/mL (116.42 mM; Need ultrasonic)
286.29
99.5%
Solid
1015474-32-4
Research
CC 122
O=C(C(N1C(C)=NC2=C(C(N)=CC=C2)C1=O)CC3)NC3=O
286.29
Research
Cancer-programmed cell death
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Avadomide, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.