missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-H7 Monoclonal Antibody (MA57YW), PE-eFluor™ 610, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 61-6537-41
Additional Details : Weight : 0.01000kg
Description
Mouse Monoclonal Antibody
B7-H7 Monoclonal antibody specifically detects B7-H7 in Human samples. It is validated for Flow CytometrySpecifications
B7-H7 | |
Monoclonal | |
5 μL/Test | |
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
Q9UM44 | |
HHLA2 | |
B7-H7 recombinant protein | |
25 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Flow Cytometry | |
MA57YW | |
PE-eFluor 610 | |
HHLA2 | |
B7-H5; B7H7; B7-H7; B7y; HERV-H LTR-associating protein 2; HHLA2; Human endogenous retrovirus-H long terminal repeat-associating protein 2 | |
Mouse | |
Affinity chromatography | |
RUO | |
11148 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |