missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-H7 Monoclonal Antibody (MA57YW), PE-eFluor™ 610, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1678.00 SEK - 3890.00 SEK
Specifications
Antigen | B7-H7 |
---|---|
Clone | MA57YW |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16371094
|
Affymetrix eBioscience
61-6537-42 |
100 Tests |
3890.00 SEK
Pack of 100 |
Estimated Shipment: 15-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16361094
|
Affymetrix eBioscience
61-6537-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Mouse Monoclonal Antibody
B7-H7 (also known as HHLA2) is a membrane protein with three Ig-like domains (IgV-IgC-IgV), whereas other members of the B7 family generally have only two Ig domains (IgV-IgC). B7-H7 is somewhat more closely related to B7-H3 and B7-H4 and shares 10% to 18% amino acid identity and 23% to 33% similarity to B7 family members. B7-H7 protein is widely expressed in epithelium cells of the gallbladder, breast, kidney, colon, small intestines and placenta. In the immune system, HHLA2 protein is not constitutively expressed on human monocytes/macrophages. B7-H7 is widely expressed in cancers, especially in triple negative breast cancer.Specifications
B7-H7 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
Q9UM44 | |
HHLA2 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
MA57YW | |
Flow Cytometry | |
PE-eFluor 610 | |
Mouse | |
Human | |
11148 | |
B7-H7 recombinant protein | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
HHLA2 |