missing translation for 'onlineSavingsMsg'
Learn More

B7-H7 Monoclonal Antibody (MA57YW), PE-eFluor™ 610, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 61-6537-42

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16371094

  • 3890.00 SEK / Pack of 100
Estimated Shipment: 15-08-2024
to see stock.

Description

Description

Mouse Monoclonal Antibody

B7-H7 (also known as HHLA2) is a membrane protein with three Ig-like domains (IgV-IgC-IgV), whereas other members of the B7 family generally have only two Ig domains (IgV-IgC). B7-H7 is somewhat more closely related to B7-H3 and B7-H4 and shares 10% to 18% amino acid identity and 23% to 33% similarity to B7 family members. B7-H7 protein is widely expressed in epithelium cells of the gallbladder, breast, kidney, colon, small intestines and placenta. In the immune system, HHLA2 protein is not constitutively expressed on human monocytes/macrophages. B7-H7 is widely expressed in cancers, especially in triple negative breast cancer.
Specifications

Specifications

B7-H7
Monoclonal
5 μL/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
Q9UM44
HHLA2
B7-H7 recombinant protein
100 Tests
Primary
Human
Antibody
IgG1 κ
Flow Cytometry
MA57YW
PE-eFluor 610
HHLA2
B7-H5; B7H7; B7-H7; B7y; HERV-H LTR-associating 2; HERV-H LTR-associating protein 2; HHLA2; Human endogenous retrovirus-H long terminal repeat-associating protein 2
Mouse
Affinity chromatography
RUO
11148
4° C, store in dark, DO NOT FREEZE!
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers