Learn More
HUBER Ball Valve
Stainless steel ball valve
Brand:  HUBER 328240
 View more versions of this product
Product Code. 15246577
Specifications
-60 to +200 deg.C; max. 10 bar at +175 deg.C
Ball Valve
M16 x 1