Learn More
HUBER Ball Valve
Stainless steel ball valve
Brand:  HUBER 328203
 View more versions of this product
Product Code. 15276537
Specifications
-60 to +200 deg.C; max. 10 bar at +175 deg.C
Ball Valve
M30 x 1.5