Learn More
VEE GEE Baume Hydrometer, Heavy Liquids
The Baume (Heavy) Hydrometer is for liquids heavier than water.
Brand: VEE GEE 6609-10
Additional Details : Weight : 0.09000kg
Description
- Specific Gravity: 145/(145-degrees Baume)
- Calibrated for use at 15.5°C/60°F
Specifications
Baume Hydrometer | |
30.5 cm | |
0 to 35° |
0.5° | |
Glass | |
15.5°C (60°F) |