Learn More
VEE GEE Baume Hydrometer, Heavy Liquids
The Baume (Heavy) Hydrometer is for liquids heavier than water.
389.00 SEK - 640.00 SEK
Specifications
Type | Baume Hydrometer |
---|---|
Length (Metric) | 30.5 cm |
Material | Glass |
TempofStandardization | 15.5°C (60°F) |
Product Code | Brand | Baume Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Baume Range | Price | Quantity & Availability | |||||
17876553
|
VEE GEE
6609-9 |
0 to 25° |
389.00 SEK
Each |
Estimated Shipment: 18-09-2024
Log in to see stock. |
|||||
17846523
|
VEE GEE
6609-10 |
0 to 35° |
389.00 SEK
Each |
Estimated Shipment: 18-09-2024
Log in to see stock. |
|||||
17866543
|
VEE GEE
6609-5 |
39 to 51° |
389.00 SEK
Each |
Estimated Shipment: 18-09-2024
Log in to see stock. |
|||||
17816553
|
VEE GEE
6609-3 |
19 to 31° |
389.00 SEK
Each |
Estimated Shipment: 18-09-2024
Log in to see stock. |
|||||
17806553
|
VEE GEE
6609-4 |
29 to 41° |
389.00 SEK
Each |
Estimated Shipment: 18-09-2024
Log in to see stock. |
|||||
17846443
|
VEE GEE
6609-14 |
0 to 70° |
389.00 SEK
Each |
Estimated Shipment: 18-09-2024
Log in to see stock. |
|||||
17896533
|
VEE GEE
6609-1 |
0 to 12° |
389.00 SEK
Each |
Estimated Shipment: 18-09-2024
Log in to see stock. |
|||||
17826473
|
VEE GEE
6609-15 |
0 to 90° |
640.00 SEK
Each |
Estimated Shipment: 11-09-2024
Log in to see stock. |
|||||
17826463
|
VEE GEE
6609-13 |
0 to 50° |
389.00 SEK
Each |
Estimated Shipment: 18-09-2024
Log in to see stock. |
|||||
17866463
|
VEE GEE
6609-12 |
35 to 70° |
389.00 SEK
Each |
Estimated Shipment: 18-09-2024
Log in to see stock. |
|||||
17806473
|
VEE GEE
6609-11 |
0 to 50° |
389.00 SEK
Each |
Estimated Shipment: 18-09-2024
Log in to see stock. |
|||||
17886443
|
VEE GEE
6609-8 |
0 to 15° |
389.00 SEK
Each |
Estimated Shipment: 18-09-2024
Log in to see stock. |
|||||
17856463
|
VEE GEE
6609-2 |
9 to 21° |
389.00 SEK
Each |
Estimated Shipment: 18-09-2024
Log in to see stock. |
|||||
Description
- Specific Gravity: 145/(145-degrees Baume)
- Calibrated for use at 15.5°C/60°F
Specifications
Baume Hydrometer | |
30.5 cm | |
Glass | |
15.5°C (60°F) |