missing translation for 'onlineSavingsMsg'
Learn More

Bcl-2 related protein A1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

4480.00 SEK

Specifications

Antigen Bcl-2 related protein A1
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18322165
View Documents
Bio-Techne
NBP3-10872-100UL
100 μg
4480.00 SEK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Bcl-2 related protein A1 Polyclonal specifically detects Bcl-2 related protein A1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Bcl-2 related protein A1
Western Blot
Unconjugated
Rabbit
Apoptosis, Cancer
PBS buffer, 2% sucrose
597
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1.0 ug/ml
Polyclonal
Purified
RUO
Human
ACC-1, ACC-2, Bcl2-L-5, BCL2L5HBPA1, Bcl-2-like protein 5, BCL2-related protein A1, BFL1bcl-2-related protein A1, GRSbcl2-L-5, hematopoietic BCL2-related protein A1, Hemopoietic-specific early response protein, Protein BFL-1, Protein GRS
The immunogen is a synthetic peptide directed towards the C terminal region of human Bcl-2 related protein A1 (NP_004040). Peptide sequence FIMNNTGEWIRQNGGWENGFVKKFEPKSGWMTFLEVTGKICEMLSLLKQY
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.