missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL2L14 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12846-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the BCL2 protein family. BCL2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. Overexpression of this gene has been shown to induce apoptosis in cells. Three alternatively spliced transcript variants encoding two distinct isoforms have been reported for this gene.Specifications
BCL2L14 | |
Polyclonal | |
Unconjugated | |
BCL2L14 | |
Apoptosis regulator Bcl G, BCL2L14 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79370 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BZR8 | |
BCL2L14 | |
BCL2L14 Fusion Protein Ag3926 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |