missing translation for 'onlineSavingsMsg'
Learn More

BCL2L14 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1656.73 SEK - 4721.67 SEK

Specifications

Antigen BCL2L14
Concentration 0.47 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16850784
View Documents Promotion Details
Proteintech
12846-1-AP-20UL
20 μL
1656.73 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16840784
View Documents Promotion Details
Proteintech
12846-1-AP-150UL
150 μL
4721.67 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The protein encoded by this gene belongs to the BCL2 protein family. BCL2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. Overexpression of this gene has been shown to induce apoptosis in cells. Three alternatively spliced transcript variants encoding two distinct isoforms have been reported for this gene.
Specifications

Specifications

BCL2L14
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Human
Q9BZR8
79370
BCL2L14 Fusion Protein Ag3926
Primary
-20°C
BCL2L14
0.47 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Apoptosis regulator Bcl G, BCL2L14
BCL2L14
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications