Learn More
BD Difco™ Listeria O Antisera
Used as positive control in identification of Listeria monocytogenes in the slide agglutination test
Brand: BD 223011
Additional Details : Weight : 0.00100kg
Description
Listeria monocytogenes is a widespread problem in public health and the food industries.
- Lyophilized, polyclonal rabbit antisera containing approximately 0.1% sodium azide as a preservative
- Listeria O Antisera Types 1 and 4 are specific for the respective serotypes of L. monocytogenes while Listeria O Antiserum Poly contains agglutinins for L. monocytogenes serotypes 1 and 4
For in vitro Diagnostic Use.
Certifications
CE marked
Specifications
CE Marked; For in vitro diagnostic use | |
Antiserum | |
1 mL |
Refrigerate, store at 2°C to 8°C | |
Used as positive control in identification of Listeria monocytogenes in the slide agglutination test |
Safety and Handling
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501-Dispose of contents/container to in accordance with all local/regional/national/internationalregulations.
Supplemental information
- MIXTURE LIST-Contains : Sodium Azide