missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Listeria O Antisera
Used as positive control in identification of Listeria monocytogenes in the slide agglutination test
1750.00 SEK - 4600.00 SEK
Specifications
Certifications/Compliance | CE Marked; For in vitro diagnostic use |
---|---|
Content And Storage | Refrigerate, store at 2°C to 8°C |
Product Type | Antiserum |
For Use With (Application) | Used as positive control in identification of Listeria monocytogenes in the slide agglutination test |
Quantity | 1 mL |
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
11712764
|
BD
223001 |
Antiserum |
4585.00 SEK
1mL |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||||
11722764
|
BD
223011 |
Antiserum |
1750.00 SEK
1mL |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
11732764
|
BD
223021 |
Antiserum |
4600.00 SEK
1mL |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
Description
Listeria monocytogenes is a widespread problem in public health and the food industries.
- Lyophilized, polyclonal rabbit antisera containing approximately 0.1% sodium azide as a preservative
- Listeria O Antisera Types 1 and 4 are specific for the respective serotypes of L. monocytogenes while Listeria O Antiserum Poly contains agglutinins for L. monocytogenes serotypes 1 and 4
For in vitro Diagnostic Use.
Certifications
CE marked
Specifications
CE Marked; For in vitro diagnostic use | |
Antiserum | |
1 mL |
Refrigerate, store at 2°C to 8°C | |
Used as positive control in identification of Listeria monocytogenes in the slide agglutination test |