Learn More
Bel-Art™ SP Scienceware™ Pipet Vacuum Aspirator Collection System
Performs vacuum aspiration of a variety of liquids, including tissue culture media, supernatants, and chemical wastes
Brand: Bel-Art™ F19917-0250
Additional Details : Weight : 2.72000kg
Description
- Built-in pump quickly aspirates fluids from pipet tip, Pasteur pipet or serological pipet inserted into the collection tube
- Maintains low noise level (76db at 1 meter) and minimal vibration
- Excellent for fluid collection from tissue and/or cell culture, Petri dishes, plates, flasks and tubes
- Chemical and corrosion resistant polypropylene collection bottle holds 3800mL (1 gallon) fluid wastes and has a safety release vent to prevent vacuum implosion
- A replaceable hydrophobic filter seals itself if it becomes wet protecting the pump motor from fluid contamination
- Lightweight and compact design will not crowd workspace in a hood or at the bench
Specifications
1.0 Gallon Collection System with Pump | |
3800 mL |