Learn More
Bel-Art™ SP Scienceware™ Pipet Vacuum Aspirator Collection System
Performs vacuum aspiration of a variety of liquids, including tissue culture media, supernatants, and chemical wastes
930.00 SEK - 5000.00 SEK
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
15237513
|
Bel-Art™
F19917-0155 |
Replacement Hydrophobic Filters |
930.00 SEK
Pack of 12 |
Estimated Shipment: 01-10-2024
Log in to see stock. |
|||||
15206327
|
Bel-Art™
F19917-0250 |
1.0 Gallon Collection System with Pump |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Built-in pump quickly aspirates fluids from pipet tip, Pasteur pipet or serological pipet inserted into the collection tube
- Maintains low noise level (76db at 1 meter) and minimal vibration
- Excellent for fluid collection from tissue and/or cell culture, Petri dishes, plates, flasks and tubes
- Chemical and corrosion resistant polypropylene collection bottle holds 3800mL (1 gallon) fluid wastes and has a safety release vent to prevent vacuum implosion
- A replaceable hydrophobic filter seals itself if it becomes wet protecting the pump motor from fluid contamination
- Lightweight and compact design will not crowd workspace in a hood or at the bench
Specifications
3800 mL |