Learn More
Kern Sohn Bench Scale ECE
Battery operation, compact, and lightweight bench scale with recessed grips on the underside.
Brand: Kern Sohn ECE 20K-2N
This item is not returnable.
View return policy
Description
- High mobility: Battery operation, compact, lightweight construction, recessed grips on the underside and low weight make this balance ideal for use in several locations
- Particularly large weighing plate
- Very fast display: steady weight values within 2 s
- Simple and convenient 2-key operation
Specifications
20 kg | |
260 x 320 mm | |
10 g |