missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Beta-Glucuronidase
BIOPHARMA

For analysis of steroid-gluceronides

Brand:  MP Biomedicals™ 0215228401

 View more versions of this product

Product Code. 11428541

  • 621.00 SEK / 1mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Beta-Glucuronidase, [9001-45-0]
  • For analysis of steroid-gluceronides
  • In 65% saturated ammonium sulfate
  • Activity: ≤5000 u/mL
  • Unit Definition: Amount that liberates 1.0μg of phenolphthalein from phenolphthalein glucuronide per hour at pH 5.0 and 37°C
TRUSTED_SUSTAINABILITY
Specifications

Specifications

beta-Glucuronidase: >50,000 U/mL, Sulfatase: 5,000 U/mL
Enzymatic hydrolysis of Glucuronides from Serum Plasma and other fluids prior to analysis by Enzyme immunoassay, mass spectrometry, GC, HPLC
Helix pomatia
beta-Glucuronidase
Solution
Brown
4°C
1 mL
D-glucuronic Acid, D-galacturonic Acid and D-glucaro-1,4-lactone
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers