missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Beta-Glucuronidase
For analysis of steroid-gluceronides
621.00 SEK - 4040.00 SEK
Specifications
Concentration | beta-Glucuronidase: >50,000 U/mL, Sulfatase: 5,000 U/mL |
---|---|
Color | Brown |
For Use With (Application) | Enzymatic hydrolysis of Glucuronides from Serum Plasma and other fluids prior to analysis by Enzyme immunoassay, mass spectrometry, GC, HPLC |
Content And Storage | 4°C |
Source | Helix pomatia |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11428541
|
MP Biomedicals™
0215228401 |
1 mL |
621.00 SEK
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11452602
|
MP Biomedicals™
0215228410 |
10 mL |
4040.00 SEK
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11438541
|
MP Biomedicals™
0215228402 |
2 mL |
1065.00 SEK
2mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Beta-Glucuronidase, [9001-45-0]
- For analysis of steroid-gluceronides.
- In 65% saturated ammonium sulfate
- Activity: ≤5000 u/mL
- Unit Definition: Amount that liberates 1.0μg of phenolphthalein from phenolphthalein glucuronide per hour at pH 5.0 and 37°C.
Specifications
beta-Glucuronidase: >50,000 U/mL, Sulfatase: 5,000 U/mL | |
Enzymatic hydrolysis of Glucuronides from Serum Plasma and other fluids prior to analysis by Enzyme immunoassay, mass spectrometry, GC, HPLC | |
Helix pomatia | |
D-glucuronic Acid, D-galacturonic Acid and D-glucaro-1,4-lactone |
Brown | |
4°C | |
beta-Glucuronidase | |
Solution |