missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Beta-Glucuronidase
For analysis of steroid-gluceronides
Brand: MP Biomedicals™ 0215228402
This item is not returnable.
View return policy
Description
Beta-Glucuronidase, [9001-45-0]- For analysis of steroid-gluceronides
- In 65% saturated ammonium sulfate
- Activity: ≤5000 u/mL
- Unit Definition: Amount that liberates 1.0μg of phenolphthalein from phenolphthalein glucuronide per hour at pH 5.0 and 37°C
Specifications
beta-Glucuronidase: >50,000 U/mL, Sulfatase: 5,000 U/mL | |
Enzymatic hydrolysis of Glucuronides from Serum Plasma and other fluids prior to analysis by Enzyme immunoassay, mass spectrometry, GC, HPLC | |
Helix pomatia | |
beta-Glucuronidase | |
Solution |
Brown | |
4°C | |
2 mL | |
D-glucuronic Acid, D-galacturonic Acid and D-glucaro-1,4-lactone |