missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Beta-Glucuronidase
For analysis of steroid-gluceronides
Brand: MP Biomedicals™ 0215228410
This item is not returnable.
View return policy
Description
Beta-Glucuronidase, [9001-45-0]- For analysis of steroid-gluceronides
- In 65% saturated ammonium sulfate
- Activity: ≤5000 u/mL
- Unit Definition: Amount that liberates 1.0μg of phenolphthalein from phenolphthalein glucuronide per hour at pH 5.0 and 37°C
Specifications
beta-Glucuronidase: >50,000 U/mL, Sulfatase: 5,000 U/mL | |
Enzymatic hydrolysis of Glucuronides from Serum Plasma and other fluids prior to analysis by Enzyme immunoassay, mass spectrometry, GC, HPLC | |
Helix pomatia | |
beta-Glucuronidase | |
Solution |
Brown | |
4°C | |
10 mL | |
D-glucuronic Acid, D-galacturonic Acid and D-glucaro-1,4-lactone |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Beta-Glucuronidase
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Ammonium Sulphate,-Glucuronidase