missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-chloroethyl) ether, 99%, Thermo Scientific Chemicals

346.00 SEK - 2725.00 SEK

Identificadores de productos químicos

CAS 111-44-4
Molecular Formula C4H8Cl2O
Molecular Weight (g/mol) 143.01
MDL Number MFCD00000975
InChI Key ZNSMNVMLTJELDZ-UHFFFAOYSA-N
Synonym 2-chloroethyl ether, 2,2'-dichlorodiethyl ether, bis 2-chloroethyl ether, chlorex, chloroethyl ether, dcee, dichloroether, khloreks, dichloroethyl ether
PubChem CID 8115
ChEBI CHEBI:34573
IUPAC Name 1-chloro-2-(2-chloroethoxy)ethane
SMILES ClCCOCCCl
Ver más especificaciones

Productos 3
Código de producto Marca Quantity Precio Cantidad y disponibilidad  
Código de producto Marca Quantity Precio Cantidad y disponibilidad  
11472347
Ver documentos
Thermo Scientific Alfa Aesar
A14234.18
50 g
346.00 SEK
50g
Please to purchase this item. Need a web account? Register with us today!
11482347
Ver documentos Detalles de la promoción
Thermo Scientific Alfa Aesar
A14234.30
250 g
845.00 SEK
250g
Please to purchase this item. Need a web account? Register with us today!
11462347
Ver documentos
Thermo Scientific Alfa Aesar
A14234.0B
1000 g
2725.00 SEK
1000g
Please to purchase this item. Need a web account? Register with us today!
Descripción

Descripción

Bis(2-chloroethyl) ether is used as a solvent and soil fumigant. It may be used in the preparation of macrocyclic polyethers, dibenzo-18-crown-6 polyether and dicyclohexyl-18-crown-6 polyether.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-chloroethyl) ether is used as a solvent and soil fumigant. It may be used in the preparation of macrocyclic polyethers, dibenzo-18-crown-6 polyether and dicyclohexyl-18-crown-6 polyether.

Solubility
Soluble in water. (1.1 g/L) at 20°C.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com
Especificaciones

Identificadores de productos químicos

111-44-4
143.01
ZNSMNVMLTJELDZ-UHFFFAOYSA-N
8115
1-chloro-2-(2-chloroethoxy)ethane
C4H8Cl2O
MFCD00000975
2-chloroethyl ether, 2,2'-dichlorodiethyl ether, bis 2-chloroethyl ether, chlorex, chloroethyl ether, dcee, dichloroether, khloreks, dichloroethyl ether
CHEBI:34573
ClCCOCCCl

Especificaciones

111-44-4
1.219
55°C (131°F)
C4H8Cl2O
1.457
UN1916
14,3066
Soluble in water. (1.1g/L) at 20°C.
ClCCOCCCl
143.01
CHEBI:34573
99%
-50°C
178°C
Pungent
ClCH2CH2OCH2CH2Cl
MFCD00000975
605317
2-chloroethyl ether, 2,2'-dichlorodiethyl ether, bis 2-chloroethyl ether, chlorex, chloroethyl ether, dcee, dichloroether, khloreks, dichloroethyl ether
ZNSMNVMLTJELDZ-UHFFFAOYSA-N
1-chloro-2-(2-chloroethoxy)ethane
8115
143.01
Bis(2-chloroethyl) ether
Videos
Seguridad y manipulación

Seguridad y manipulación

GHS H Statement
H300-H310-H330-H226-H351
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Flammable liquid and vapour.
Suspected of causing cancer.

P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P310-P330-P363-P370+P378q-P501c

H226-H300+H310+H330

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 2,2'-DICHLORODIETHYL ETHER

missing translation for 'einecsNumber' : 203-870-1

missing translation for 'rtecsNumber' : KN0875000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documentos

Documentos

Product Certifications

RUO – Research Use Only