missing translation for 'onlineSavingsMsg'
Learn More

BJHCC20A Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

3675.00 SEK - 5775.00 SEK

Specifications

Antigen BJHCC20A
Dilution Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18321507
View Documents
Bio-Techne
NBP3-17470-25UL
25 μg
3675.00 SEK
25µL
Please to purchase this item. Need a web account? Register with us today!
18337244
View Documents
Bio-Techne
NBP3-17470-100UL
100 μg
5775.00 SEK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

BJHCC20A Polyclonal antibody specifically detects BJHCC20A in Human samples. It is validated for Immunofluorescence
Specifications

Specifications

BJHCC20A
Immunofluorescence
Unconjugated
Rabbit
Human
BJHCC20A, Cancer/testis antigen 55, chromosome X open reading frame 48, CT55, CT55 BRCA2-interacting protein, CXorf48, FLJ20527, RP13-565O16.1, Tumor antigen BJ-HCC-20
This antibody was developed against Recombinant Protein corresponding to amino acids: GNVPLKVGQKVNVVVEEDKPHYGLRAIKVDVVPRHLYGAGPSDSGTRVLIG
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL
Polyclonal
Purified
RUO
PBS, pH 7.2, 40% glycerol
54967
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.