Learn More
Blarcamesine hydrochloride, MedChemExpress
MedChemExpress Blarcamesine hydrochloride is a Sigma-1 Receptor agonist with an IC50 of 860 nM.
Brand: Medchem Express HY-101864
This item is not returnable.
View return policy
Description
Blarcamesine hydrochloride is a Sigma-1 Receptor agonist with an IC50 of 860 nM.Specifications
Blarcamesine hydrochloride | |
White | |
C19H24ClNO | |
H2O : 100 mg/mL (314.61 mM; Need ultrasonic) ∣DMSO : 25 mg/mL (78.65 mM; Need ultrasonic) | |
317.85 | |
99.85% | |
Solid |
195615-84-0 | |
Research | |
5 mg | |
CN(C)CC1CCOC1(C2=CC=CC=C2)C3=CC=CC=C3.[H]Cl | |
317.85 | |
Research | |
Neuroscience-Neurodegeneration |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.