Learn More
Blarcamesine hydrochloride, MedChemExpress
MedChemExpress Blarcamesine hydrochloride is a Sigma-1 Receptor agonist with an IC50 of 860 nM.
Brand:  Medchem Express HY-101864
Encompass_Preferred
 View more versions of this product
Product Code. 16466347
  • 1609.28 SEK / 5mg
Explore more special offers
Description
Description
Blarcamesine hydrochloride is a Sigma-1 Receptor agonist with an IC50 of 860 nM.
Specifications
Specifications
Blarcamesine hydrochloride
White
C19H24ClNO
H2O : 100 mg/mL (314.61 mM; Need ultrasonic) ∣DMSO : 25 mg/mL (78.65 mM; Need ultrasonic)
317.85
99.85%
Solid
195615-84-0
Research
5 mg
CN(C)CC1CCOC1(C2=CC=CC=C2)C3=CC=CC=C3.[H]Cl
317.85
Research
Neuroscience-Neurodegeneration
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.