Learn More
Blarcamesine hydrochloride, MedChemExpress
MedChemExpress Blarcamesine hydrochloride is a Sigma-1 Receptor agonist with an IC50 of 860 nM.
1609.28 SEK - 14841.13 SEK
Specifications
Chemical Name or Material | Blarcamesine hydrochloride |
---|---|
CAS | 195615-84-0 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C19H24ClNO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16493618
|
Medchem Express
HY-101864 |
10 mg |
2741.74 SEK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16474706
|
Medchem Express
HY-101864 |
100 mg |
14841.13 SEK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16466347
|
Medchem Express
HY-101864 |
5 mg |
1609.28 SEK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16430017
|
Medchem Express
HY-101864 |
50 mg |
9521.58 SEK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Blarcamesine hydrochloride is a Sigma-1 Receptor agonist with an IC50 of 860 nM.Specifications
Blarcamesine hydrochloride | |
White | |
C19H24ClNO | |
CN(C)CC1CCOC1(C2=CC=CC=C2)C3=CC=CC=C3.[H]Cl | |
317.85 | |
Research | |
Neuroscience-Neurodegeneration |
195615-84-0 | |
Research | |
H2O : 100 mg/mL (314.61 mM; Need ultrasonic) ∣DMSO : 25 mg/mL (78.65 mM; Need ultrasonic) | |
317.85 | |
99.85% | |
Solid |
Safety and Handling
H315∣H319∣H335
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.