Learn More
Brady™ BMP61 BMP71 M611 Polyester Labels
Terminal block markers for the BMP61 and M611 Thermal Transfer Portable Printers.
Brand:  Brady™ 217003
 View more versions of this product
Product Code. 11528624
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Material is UL approved with its respective ribbon
  • See the Recommended Ribbon Series for the appropriate ribbon to use
  • Can also be printed on the BMP™71 Label Printer provided the roll adapter (M71-ADAPT) is used
Specifications
Specifications
Permanent Acrylic
Gloss
Thermal Transfer
15 m x 8 mm
White
Polyester (B-422)
BMP61, BMP71, M611