Learn More
BMS 777607, MedChemExpress
MedChemExpress BMS 777607 (BMS 817378) is a Met-related inhibitor for c-Met, Axl, Ron and Tyro3 with IC50s of 3.9 nM, 1.1 nM, 1.8 nM and 4.3 nM, respectively, and 40-fold more selective for Met-related targets than Lck, VEGFR-2, and TrkA/B, with more than 500-fold greater selectivity versus all other receptor and non receptor kinases.
Brand:  Medchem Express HY-12076
Encompass_Preferred
 View more versions of this product
Product Code. 16454978
  • 2536.38 SEK / 10mg
Explore more special offers
Description
Description
BMS 777607 (BMS 817378) is a Met-related inhibitor for c-Met, Axl, Ron and Tyro3 with IC50s of 3.9 nM, 1.1 nM, 1.8 nM and 4.3 nM, respectively, and 40-fold more selective for Met-related targets than Lck, VEGFR-2, and TrkA/B, with more than 500-fold greater selectivity versus all other receptor and non receptor kinases.
Specifications
Specifications
BMS 777607
White
C25H19ClF2N4O4
BMS 817378
O=C(NC1=CC=C(C(F)=C1)OC2=C(C(N)=NC=C2)Cl)C3=C(C=CN(C3=O)C4=CC=C(C=C4)F)OCC
512.89
Research
Cancer-Kinase/protease
1025720-94-8
Research
10 mg
DMSO : ≥ 39 mg/mL (76.04 mM)
512.89
98.49%
Solid
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
BMS 777607, MedChemExpress