Learn More
BMS 777607, MedChemExpress
MedChemExpress BMS 777607 (BMS 817378) is a Met-related inhibitor for c-Met, Axl, Ron and Tyro3 with IC50s of 3.9 nM, 1.1 nM, 1.8 nM and 4.3 nM, respectively, and 40-fold more selective for Met-related targets than Lck, VEGFR-2, and TrkA/B, with more than 500-fold greater selectivity versus all other receptor and non receptor kinases.
Brand: Medchem Express HY-12076
Description
BMS 777607 (BMS 817378) is a Met-related inhibitor for c-Met, Axl, Ron and Tyro3 with IC50s of 3.9 nM, 1.1 nM, 1.8 nM and 4.3 nM, respectively, and 40-fold more selective for Met-related targets than Lck, VEGFR-2, and TrkA/B, with more than 500-fold greater selectivity versus all other receptor and non receptor kinases.Specifications
BMS 777607 | |
White | |
C25H19ClF2N4O4 | |
BMS 817378 | |
O=C(NC1=CC=C(C(F)=C1)OC2=C(C(N)=NC=C2)Cl)C3=C(C=CN(C3=O)C4=CC=C(C=C4)F)OCC | |
512.89 | |
Research | |
Cancer-Kinase/protease |
1025720-94-8 | |
Research | |
10 mg | |
DMSO : ≥ 39 mg/mL (76.04 mM) | |
512.89 | |
98.49% | |
Solid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only