Learn More
Bohlender™ BOLA™ Flat Flange Lids
Flat Flange Lids suitable for reaction vessels with SCHOTT flat flange.
Brand: Bohlender™ B210-12
This item is not returnable.
View return policy
Description
- Suitable for reaction vessels with SCHOTT flat flange
- Center neck socket NS 29/32
- Lateral necks with ground joint sockets or GL-thread
- Bottom side with centring collar for non-slipping alignment on the vessel
- All lateral connectors are made to insert tubing, tubes and probes either aside the center or vertical (parallel to the stirrer shaft) in order to avoid collisions
Specifications
PTFE | |
Reaction Vessels with SCHOTT Flat Flange | |
DN 100 | |
Lateral Necks: 2 x NS 29/32 Angled Aside the Shaft, 1 x NS 19/26 and 3 x GL 25 Vertical |