missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bohlender™ BOLA™ Flat Flange Lids
Flat Flange Lids suitable for reaction vessels with SCHOTT flat flange.
5681.60 SEK - 11490.40 SEK
Specifications
Material | PTFE |
---|---|
For Use With (Equipment) | Reaction Vessels with SCHOTT Flat Flange |
Product Code | Brand | Joint Type | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Joint Type | Size | Price | Quantity & Availability | ||||
17136964
|
Bohlender™
B210-02 |
DN 60 | Lateral Necks: 2 x NS 14/23 and 2 x GL 18 Angled Aside the Shaft |
5681.60 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17146964
|
Bohlender™
B210-12 |
DN 100 | Lateral Necks: 2 x NS 29/32 Angled Aside the Shaft, 1 x NS 19/26 and 3 x GL 25 Vertical |
7992.40 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17156964
|
Bohlender™
B210-14 |
DN 100 | Lateral Necks: 3 x NS 29/32 Angled Aside the Shaft |
7674.40 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17166964
|
Bohlender™
B210-22 |
DN 150 | Lateral Necks: 2 x NS 29/32, 1 x NS 19/26 and 4 x GL 25 Vertical |
9868.60 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17176964
|
Bohlender™
B210-24 |
DN 150 | Lateral Necks: 3 x NS 29/32 Angled Aside the Shaft |
11490.40 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Suitable for reaction vessels with SCHOTT flat flange
- Center neck socket NS 29/32
- Lateral necks with ground joint sockets or GL-thread
- Bottom side with centring collar for non-slipping alignment on the vessel
- All lateral connectors are made to insert tubing, tubes and probes either aside the center or vertical (parallel to the stirrer shaft) in order to avoid collisions
Specifications
PTFE | |
Reaction Vessels with SCHOTT Flat Flange |