missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Borate-buffered saline (5X)

Brand:  Thermo Scientific™ J60979.AP

 View more versions of this product

Product Code. 15423659

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Borate buffered saline has been used to determine whether heat-killed, borate-buffered suspensions of Gordonia bronchialis, Rhodococcus coprophilus or Tsukamurella inchonensis can be used to treat canine flea allergy. Borate buffered saline has also been used to investigate the effects of lentivirus mediated siRNA targeting human metastasis suppressor 1 (MTSS1, MIM-B gene) gene on the invasive and metastatic potentials of hepatocellular carcinoma (HCC) MHCC97H cells. Borate buffered saline has been used to determine the effects of packaging solution osmolality and buffering agent on soft contact lens parameters. Borate buffer solutions is used in protein modification procedures requiring amine-free buffer at alkaline pH.

Solubility
Soluble in water 0.1 M at 20°C.

Notes
125mM sodium borate, 750mM sodium chloride, pH 7.0
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Borate-buffered Saline
Liquid
Ambient temperatures
MFCD00081185
[Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
201.21
500 mL
5X
B4Na2O7
NXSVMNYRVBQCMO-UHFFFAOYSA-N
disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers