missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Borate-buffered saline (5X)

669.00 SEK - 795.00 SEK

Specifications

Product Type Borate-buffered Saline
Form Liquid
Concentration 5X
Content And Storage Ambient temperatures
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15413659
missing translation for 'viewDocuments'
Thermo Scientific™
J60979.AK
250 mL
669.00 SEK
250mL
Please to purchase this item. Need a web account? Register with us today!
15423659
missing translation for 'viewDocuments'
Thermo Scientific™
J60979.AP
500 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Borate buffered saline has been used to determine whether heat-killed, borate-buffered suspensions of Gordonia bronchialis, Rhodococcus coprophilus or Tsukamurella inchonensis can be used to treat canine flea allergy. Borate buffered saline has also been used to investigate the effects of lentivirus mediated siRNA targeting human metastasis suppressor 1 (MTSS1, MIM-B gene) gene on the invasive and metastatic potentials of hepatocellular carcinoma (HCC) MHCC97H cells. Borate buffered saline has been used to determine the effects of packaging solution osmolality and buffering agent on soft contact lens parameters. Borate buffer solutions is used in protein modification procedures requiring amine-free buffer at alkaline pH.

Solubility
Soluble in water 0.1 M at 20°C.

Notes
125mM sodium borate, 750mM sodium chloride, pH 7.0
Specifications

Specifications

Borate-buffered Saline
Liquid
5X
Ambient temperatures
Videos
SDS
Documents

Documents

Certificates