Learn More
BRAND™ BLAUBRAND™ Borosilicate Glass Centrifuge Tube
Withstands RCF up to 700. Design and accuracy meet ASTM (American Society for Testing and Materials) specifications.
Brand:  BRAND™ 362138
1619.19 SEK valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11884050
  • 1865.00 SEK / Pack of 2
Estimated Shipment: 26-08-2024
to see stock.
Add to basket
Description
Description
  • Withstands RCF up to 700.
  • Borosilicate glass construction (Boro 3.3)
  • Cylindrical with conical bottom
Specifications
Specifications
100 mL
160 mm
Conical
700 x g