Learn More
Soda Lime Glass Petri Dishes
High-quality petri dish, with lid.
Brand: Fisherbrand 2290/150X25
Description
- Constructed of soda lime glass.
- Includes lid
Specifications
Closure Included | |
Round | |
25 mm |
Soda Lime Glass | |
Petri Dish | |
150 mm |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Soda Lime Glass Petri Dishes > Dimensions: 150 dia. x 25mmH
Spot an opportunity for improvement?Share a Content Correction