Learn More
C3/C3b/C3c Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21337-1-AP-150UL
 View more versions of this product
Product Code. 16840945
  • 4652.90 SEK / 150µL
Explore more special offers
This item is not returnable. View return policy
Description
Description
This antibody raised against 1314-1663 aa of human C3 protein recognize C3 alpha chain, C3b alpha' chain, and C3c alpha' chain fragment 2.
Complement component C3 plays a central role in the activation of complement system. Its activation is required for both classical and alternative complement activation pathways. People with C3 deficiency are susceptible to bacterial infection.
Specifications
Specifications
C3/C3b/C3c
Polyclonal
Unconjugated
C3
ARMD9, ASP, C3, C3b, C3c, Complement C3, complement component 3, CPAMD1
Rabbit
Antigen Affinity Chromatography
RUO
24232, 718
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P01024, P01026
C3
C3/C3b/C3c Fusion Protein Ag15537
150 μL
Primary
Human, Rat