missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C3/C3b/C3c Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | C3/C3b/C3c |
---|---|
Concentration | 0.3 mg/mL |
Applications | Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16850945
|
Proteintech
21337-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16840945
|
Proteintech
21337-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody raised against 1314-1663 aa of human C3 protein recognize C3 alpha chain, C3b alpha' chain, and C3c alpha' chain fragment 2.
Complement component C3 plays a central role in the activation of complement system. Its activation is required for both classical and alternative complement activation pathways. People with C3 deficiency are susceptible to bacterial infection.Spécification
C3/C3b/C3c | |
Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Rat, Human | |
P01024, P01026 | |
24232, 718 | |
C3/C3b/C3c Fusion Protein Ag15537 | |
Primary | |
-20°C | |
C3 |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ARMD9, ASP, C3, C3b, C3c, Complement C3, complement component 3, CPAMD1 | |
C3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |