Learn More
Hamilton™ Cable
Brand:  Hamilton™ 355045
 View more versions of this product
Product Code. 10179540
  • 530.00 SEK / Each
Estimated Shipment: 29-04-2025
to see stock.
Explore more special offers
Specifications
DIN
Cable
1 mm
Product Title
Hamilton™ Cable > Length: 1mm