Learn More
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
11527874
|
Hamilton™
355158 |
2 m |
1360.00 SEK
Each |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||||
10179540
|
Hamilton™
355045 |
1 mm |
530.00 SEK
Each |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||||
Specifications
DIN | |
Cable |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Hamilton™ Cable