Learn More
Hamilton™ Cable
Brand:  Hamilton™ 355158
 View more versions of this product
Product Code. 11527874
  • 1360.00 SEK / Each
Estimated Shipment: 29-04-2025
to see stock.
Explore more special offers
Specifications
DIN
Cable
2 m
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number185071 Certificate TypeCertificate of Compliance Date04/03/2025 Catalog Number355158
Lot Number184546 Certificate TypeCertificate of Compliance Date21/01/2025 Catalog Number355158
Product Title
Hamilton™ Cable > Length: 2m