Learn More
Carbendazim, MedChemExpress
MedChemExpress Carbendazim is a potent and orally active broad-spectrum benzimidazole fungicide and can be acts as a pesticide for fungal diseases research, such as Seproria, Fusarium and Sclerotina. Carbendazim is a benzimidazole (HY-Y1825) derivative with antitumor activity and used for cancer research, especially advanced solid tumors and lymphoma.
Brand: Medchem Express HY-13582
Description
Carbendazim is a potent and orally active broad-spectrum benzimidazole fungicide and can be acts as a pesticide for fungal diseases research, such as Seproria, Fusarium and Sclerotina. Carbendazim is a benzimidazole (HY-Y1825) derivative with antitumor activity and used for cancer research, especially advanced solid tumors and lymphoma.Specifications
Carbendazim | |
10605-21-7 | |
500 mg | |
C9H9N3O2 | |
DMSO : 6.8 mg/mL (35.57 mM; Need ultrasonic) | |
191.19 | |
99.81% |
Solid | |
White | |
Research | |
COVID-19-immunoregulation | |
O=C(OC)NC1=NC2=CC=CC=C2N1 | |
191.19 | |
Research |
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only