Learn More
Carbendazim, MedChemExpress
MedChemExpress Carbendazim is a potent and orally active broad-spectrum benzimidazole fungicide and can be acts as a pesticide for fungal diseases research, such as Seproria, Fusarium and Sclerotina. Carbendazim is a benzimidazole (HY-Y1825) derivative with antitumor activity and used for cancer research, especially advanced solid tumors and lymphoma.

482.38 SEK - 5212.81 SEK
Specifications
Chemical Name or Material | Carbendazim |
---|---|
Physical Form | Solid |
CAS | 10605-21-7 |
Color | White |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16471359
![]() |
Medchem Express
HY-13582 |
1 g |
1742.78 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16466236
![]() |
Medchem Express
HY-13582 |
100 mg |
482.38 SEK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16451676
![]() |
Medchem Express
HY-13582 |
500 mg |
1198.17 SEK
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16428447
![]() |
Medchem Express
HY-13582 |
5 g |
5212.81 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Carbendazim is a potent and orally active broad-spectrum benzimidazole fungicide and can be acts as a pesticide for fungal diseases research, such as Seproria, Fusarium and Sclerotina. Carbendazim is a benzimidazole (HY-Y1825) derivative with antitumor activity and used for cancer research, especially advanced solid tumors and lymphoma.Specifications
Carbendazim | |
10605-21-7 | |
Research | |
COVID-19-immunoregulation | |
O=C(OC)NC1=NC2=CC=CC=C2N1 | |
191.19 | |
Research |
Solid | |
White | |
C9H9N3O2 | |
DMSO : 6.8 mg/mL (35.57 mM; Need ultrasonic) | |
191.19 | |
99.81% |
Safety and Handling
H340∣H360
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.