missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD278 (ICOS) Monoclonal Antibody (ISA-3), Brilliant Ultra Violet™ 661, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 376-9948-42
2456.78 SEK valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
ICOS (CD278) encodes a protein that is a member of the CD28 and CTLA-4 cell-surface receptor family. It functions as a homodimer and has a crucial role in cell-cell signaling, immune responses, and the regulation of cell proliferation. ICOS has been linked to various disorders, including Immunodeficiency, Common Variable, 1 and Common Variable Immunodeficiency.
Specifications
CD278 (ICOS) | |
Monoclonal | |
5 μL/Test | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Q9Y6W8 | |
ICOS | |
Affinity chromatography | |
RUO | |
29851 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
ISA-3 | |
Brilliant Ultraviolet 661 | |
ICOS | |
Activation-inducible lymphocyte immunomediatory molecule; Ailim; CCLP; CD278; CD28 and CTLA-4-like protein; CD28-related protein 1; CRP-1; CVID1; H4; Icos; inducible costimulator; inducible T cell costimulator; inducible T cell co-stimulator; inducible T-cell costimulator; inducible T-cell co-stimulator; Ly115 | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD278 (ICOS) Monoclonal Antibody (ISA-3), Brilliant Ultra Violet™ 661, eBioscience™, Invitrogen™ > 100 Tests; Brilliant Ultra Violet 661
Spot an opportunity for improvement?Share a Content Correction