missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD278 (ICOS) Monoclonal Antibody (ISA-3), Brilliant Ultra Violet™ 661, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1740.00 SEK - 3560.00 SEK
Specifications
Antigen | CD278 (ICOS) |
---|---|
Clone | ISA-3 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17887090
|
Invitrogen
376-9948-41 |
25 Tests |
1740.00 SEK
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17897090
|
Invitrogen
376-9948-42 |
100 Tests |
3560.00 SEK
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ICOS (CD278) encodes a protein that is a member of the CD28 and CTLA-4 cell-surface receptor family. It functions as a homodimer and has a crucial role in cell-cell signaling, immune responses, and the regulation of cell proliferation. ICOS has been linked to various disorders, including Immunodeficiency, Common Variable, 1 and Common Variable Immunodeficiency.Specifications
CD278 (ICOS) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Activation-inducible lymphocyte immunomediatory molecule; Ailim; CCLP; CD278; CD28 and CTLA-4-like protein; CD28-related protein 1; CRP-1; CVID1; H4; Icos; inducible costimulator; inducible T cell costimulator; inducible T cell co-stimulator; inducible T-cell costimulator; inducible T-cell co-stimulator; Ly115 | |
ICOS | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
ICOS |
ISA-3 | |
Flow Cytometry | |
Brilliant Ultraviolet 661 | |
Mouse | |
Human | |
Q9Y6W8 | |
29851 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |