missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD33 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17425-1-AP-20UL
This item is not returnable.
View return policy
Description
CD33 is a transmembrane protein of the sialic acid-binding immunoglobulin-like lectin (Siglec) family. It belongs to the immunoreceptor tyrosine-based inhibitory motif (ITIM)-containing molecules able of recruiting protein tyrosine phosphatases SHP-1 and SHP-2 to signal assemblies, and these ITIMs are also used for ubiquitin-mediated removal of the receptor from the cell surface. CD33 is expressed on cells of myelomonocytic lineage, binds sialic acid residues in N- and O-glycans on cell surfaces, and is a therapeutic target for acute myeloid leukemia. Further, CD33 is found on granulocyte and macrophage precursors in the bone marrow, but is not on pluripotent stem cells. CD33 is also expressed on, and is a useful marker for, peripheral monocytes. CD33 is useful for distinguishing myelogenous leukemia cells from lymphoid or erythroid leukemias. Diseases associated with CD43 dysfunction include gallbladder lymphoma and extracutaneous mastocytoma.Specifications
CD33 | |
Polyclonal | |
Unconjugated | |
CD33 | |
CD33, CD33 molecule, gp67, p67, SIGLEC 3, SIGLEC3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
945 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P20138 | |
CD33 | |
CD33 Fusion Protein Ag11490 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction