missing translation for 'onlineSavingsMsg'
Learn More

CD71 / Transferrin Receptor Mouse, Clone: TFRC/1818, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 7037-MSM8-P1

 View more versions of this product

Product Code. 15910948

  • 5965.00 SEK / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

CD71 / Transferrin Receptor
Cell surface
TFRC/1818
Unconjugated
NP_001300894
Mouse
85-95kDa (monomer) ;190kDa (dimer)
Protein A/G Purified
RUO
7037
IgG2b κ
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
TFRC
Recombinant human TFRC protein fragment (aa94-212)
PC3 cells. Human placenta or breast carcinoma.
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
CD71 / Transferrin Receptor Mouse, Clone: TFRC/1818, enQuireBio™ > 100 μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.