missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD71 / Transferrin Receptor Mouse, Clone: TFRC/1818, enQuireBio™
Mouse Monoclonal Antibody
2050.00 SEK - 5965.00 SEK
Specifications
Antigen | CD71 / Transferrin Receptor |
---|---|
Clone | TFRC/1818 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Specifications
CD71 / Transferrin Receptor | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
7037 | |
Recombinant human TFRC protein fragment (aa94-212) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
85-95kDa (monomer) ;190kDa (dimer) |
TFRC/1818 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_001300894 | |
TFRC | |
IgG2b κ | |
Protein A/G Purified | |
Cell surface | |
PC3 cells. Human placenta or breast carcinoma. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD71 / Transferrin Receptor Mouse, Clone: TFRC/1818, enQuireBio™