Learn More
Eurofins DiscoverX Cdk7/Cyclin H/MAT1 (CAK complex) Protein, Active
Active complex of recombinant full length human Cdk7, containing a C-terminal His6-tag, recombinant full-length human Cyclin H untagged and recombinant full-length human MAT1 containing an N-terminal GST-tag, for use in Kinase Assays.
5796.39 SEK - 44240.30 SEK
Specifications
Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
---|---|
Molecular Weight (g/mol) | 39.9 kDa (CDK7), 37.8 kDa (Cyclin H) and 62.8 kDa (MAT1) |
Species | Human |
Recombinant | Recombinant |
Protein Tag | C-terminal 6His / untagged / N-terminal GST |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17300224
|
Eurofins DiscoverX
14-476 |
10 μg |
5796.39 SEK
10µg |
Estimated Shipment: 08-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17310224
|
Eurofins DiscoverX
14-476M |
250 μg |
44240.30 SEK
250µg |
Estimated Shipment: 02-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Cyclin-dependent Kinase 7 (cdk7) is a member of the cyclin-dependent protein kinase (CDK) family. CDK family members are highly similar to the gene products of Saccharomyces cerevisiae cdc28, and Schizosaccharomyces pombe cdc2, and are known to be important regulators of cell cycle progression. This protein forms a trimeric complex with cyclin H and MAT1, which functions as a Cdk-activating kinase (CAK). It is an essential component of the transcription factor TFIIH, that is involved in transcription initiation and DNA repair. This protein is thought to serve as a direct link between the regulation of transcription and the cell cycle.
GenBank X79193 / GenBank U11791 / GenBank X87843
Specifications
mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. | |
Human | |
C-terminal 6His / untagged / N-terminal GST | |
Full-length | |
CDK7/Cyclin E/MAT1, active |
39.9 kDa (CDK7), 37.8 kDa (Cyclin H) and 62.8 kDa (MAT1) | |
Recombinant | |
Recombinant enzyme expressed in Sf21 insect cells | |
Combined ≥54% |