missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cefadroxil, MP Biomedicals™
816.20 SEK - 2517.50 SEK
Chemical Identifiers
CAS | 50370-12-2 |
---|---|
Molecular Formula | C16H17N3O5S |
Molecular Weight (g/mol) | 363.388 |
MDL Number | MFCD00865091 |
InChI Key | BOEGTKLJZSQCCD-UEKVPHQBSA-N |
Synonym | p-hydroxycephalexine, Cefamox |
PubChem CID | 47965 |
ChEBI | CHEBI:3479 |
IUPAC Name | (6R,7R)-7-[[(2R)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid |
SMILES | CC1=C(N2C(C(C2=O)NC(=O)C(C3=CC=C(C=C3)O)N)SC1)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11427881
|
MP Biomedicals
0215058101 |
1 g |
816.20 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11437881
|
MP Biomedicals
0215058105 |
5 g |
2517.50 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
50370-12-2 | |
363.388 | |
BOEGTKLJZSQCCD-UEKVPHQBSA-N | |
47965 | |
(6R,7R)-7-[[(2R)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid |
C16H17N3O5S | |
MFCD00865091 | |
p-hydroxycephalexine, Cefamox | |
CHEBI:3479 | |
CC1=C(N2C(C(C2=O)NC(=O)C(C3=CC=C(C=C3)O)N)SC1)C(=O)O |
Specifications
Cefadroxil | |
50370-12-2 | |
MFCD00865091 | |
BOEGTKLJZSQCCD-UEKVPHQBSA-N | |
(6R,7R)-7-[[(2R)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid | |
47965 | |
363.4 | |
Cefadroxil |
Crystalline Powder | |
C16H17N3O5S | |
p-hydroxycephalexine, Cefamox | |
CC1=C(N2C(C(C2=O)NC(=O)C(C3=CC=C(C=C3)O)N)SC1)C(=O)O | |
363.388 | |
CHEBI:3479 | |
White |